Order Entry
France
Orders LinkContactUsLinkComponent
Benzophenone

Benzophenone

Fiber drum, 25 kg
Catalog # SPCMB1088-25KGBL
Environment
Supplier:   Spectrum Chemical
Benzophenone
Catalog # SPCMB1088-25KGBL
Supplier:  Spectrum Chemical
CAS Number:  119-61-9
Benzophenone

Fiber drum, 25 kg

CAS Number:  119-61-9
Price
Inquire For Price
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Diphenylmethanone, Diphenyl ketone
  • CAS Number:119-61-9
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00003076
  • Molecular Weight:182.22 g/mol
  • Merck Index:13,01099
  • Boiling Pt:305.4 °C (1013 hPa)
  • Melting Pt:47…49 °C
  • Density:1.085 g/cm³ (20 °C)
  • Flash Pt:150 °C
  • Storage Temperature:Ambient
  • EINECS:204-337-6
  • UN:3077
  • ADR:9,III
Specifications

Specification Test Results

  • Assay:
    99% Min.

Specifications

  • Pack type:
    Fiber drum
  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.