2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride
CAS Number:86604-75-3
:
Formula:C₉H₁₂ClNO
MDL Number:MFCD00277472
:222.11 g/mol
Melting Pt:∼ 128 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together