Order Entry
France
ContactUsLinkComponent
Gold 95% supplied in 0.1 mM PBS, nanoparticles 100 nm OD1 (Abs. 572 nm)
Gold 95% supplied in 0.1 mM PBS, nanoparticles 100 nm OD1 (Abs. 572 nm)
Catalog # J67348.AC
CAS Number:  
Gold 95% supplied in 0.1 mM PBS, nanoparticles 100 nm OD1 (Abs. 572 nm)
Catalog # J67348.AC
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

About this item

 :
 :196.97 g/mol
Storage Temperature:Refrigerator
CAS Number:7440-57-5
Formula:Au
MDL Number:MFCD00003436