Order Entry
France
ContactUsLinkComponent
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 60 nm OD1 (Abs. 540 nm)
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 60 nm OD1 (Abs. 540 nm)
Catalog # J67306.AC
CAS Number:  
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 60 nm OD1 (Abs. 540 nm)
Catalog # J67306.AC
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

About this item

 :
 :196.97 g/mol
Storage Temperature:Refrigerator
CAS Number:7440-57-5
Formula:Au
MDL Number:MFCD00003436