Order Entry
France
ContactUsLinkComponent
Riboflavine (vitamin B2) 97% for electrophoresis
Riboflavine (vitamin B2) 97% for electrophoresis
Catalog # J66632.14
CAS Number:  
Riboflavine (vitamin B2) 97% for electrophoresis
Catalog # J66632.14
CAS Number:  

Specifications

  • Pk:
    25 g

Specifications

About this item

 :
 :376.37 g/mol
Merck Index:14,8200
Melting Pt:290 °C
Storage Temperature:Refrigerator
EINECS:201-507-1
CAS Number:83-88-5
Formula:C₁₇H₂₀N₄O₆
MDL Number:MFCD00005022