Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Z-Asp(OMe)-OH
Z-Asp(OMe)-OH 98%
Z-Asp(OMe)-OH 98%
Z-Asp(OMe)-OH 98%
H63642.06
:
Thermo Fisher Scientific
:
Z-Asp(OMe)-OH 98%
H63642.06
:
Thermo Fisher Scientific
:
Pk:
5 g
:
3160-47-2
:
:
C₁₃H₁₅NO₆
:
MFCD00133585
:
281.264 g/mol