

Propanephosphonic acid cyclic anhydride ≥50% (w/w) in dichloromethane
Catalog # H63583.30
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:250 g
Specifications
About this item
:
:318.19 g/mol
Storage Temperature:Ambient
EINECS:422-210-5
UN:2922
ADR:8,III
:GHS05
:GHS08
CAS Number:68957-94-8
Formula:C₉H₂₁O₆P₃
MDL Number:MFCD00006583