1-(Mesitylene-2-sulphonyl)-3-nitro-1,2,4-triazole ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-(Mesitylsulphonyl)-3-nitro-1H-1,2,4-triazole
CAS Number:74257-00-4
:
Formula:C₁₁H₁₂N₄O₄S
MDL Number:MFCD00009754
:296.301 g/mol
Melting Pt:134…139 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together