N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:152120-61-1
:
Formula:C₉H₁₄N₄O₂
MDL Number:MFCD00216663
:210.24 g/mol
Melting Pt:97…101 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together