
(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96443-42-4
:
Formula:C₈H₁₂O₄
MDL Number:MFCD04972280
:172.18 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together