Order Entry
France
ContactUsLinkComponent
Neodymium ≥99.8% (REO, rare earth oxide basis), powder -40 mesh
Flammable
Neodymium ≥99.8% (REO, rare earth oxide basis), powder -40 mesh
Catalog # 44181.06
CAS Number:  
Flammable
Neodymium ≥99.8% (REO, rare earth oxide basis), powder -40 mesh
Catalog # 44181.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :144.24 g/mol
Merck Index:12,06538
Boiling Pt:2474 °C (1013 hPa)
Melting Pt:1014.85 °C
Density:6.46 g/cm³ (1240 °C)
Storage Temperature:Ambient
EINECS:231-109-3
UN:3089
ADR:4.1,II
 :ampouled under argon
 :GHS02
CAS Number:7440-00-8
Formula:Nd
MDL Number:MFCD00011130