4-Methyl-1,2-cyclohexene oxide cis- and trans-mixture 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36099-51-1
:
Formula:C₇H₁₂O
MDL Number:MFCD09742280
:112.17 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together