
1,2,3,4-Tetrahydro-2,2,4,7-tetramethylquinoline 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59388-58-8
:
Formula:C₁₃H₁₉N
MDL Number:MFCD00799587
:189.30 g/mol
Boiling Pt:123 °C (5 mmHg)
Density:0.975
Flash Pt:>110 °C (230 °F)
EINECS:261-732-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together