(R)-(+)-2-(Boc-Aminomethyl)-4,4-diethoxypiperidine fumarate
Catalog # H25909.ME
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
Formula:C₁₅H₃₀N₂O₄·C₄H₄O₄
MDL Number:MFCD09953465
:418.49 g/mol
:GHS07
Storage Temperature:Ambient