Morpholine-4-carboxamidine hydrobromide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:157415-17-3
:
Formula:C₅H₁₂BrN₃O
MDL Number:MFCD00051994
:210.075 g/mol
Melting Pt:∼ 155…165 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together