
cis-Aconitic anhydride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6318-55-4
:
Formula:C₆H₄O₅
MDL Number:MFCD00005524
:156.093 g/mol
Melting Pt:74…78 °C
EINECS:228-663-3
ADR:8,III
:GHS05
Related Information
Frequently Bought Together