Triethyl-4-phosphonocrotonate cis- and trans-mixture 94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10236-14-3
:
Formula:C₁₀H₁₉O₅P
MDL Number:MFCD00009192
:250.231 g/mol
Boiling Pt:133…135 °C (0.4 mmHg)
Density:1.130
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together