Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers), tech.
: Methylnadic Anhydride, Methylnadic anhydride, Nadic Methyl Anhydride, Methyl-3,6-endomethylenetetrahydrophthalic Anhydride
:25134-21-8
:
:C₁₀H₁₀O₃
:MFCD00167036
:178.19 g/mol
:137…140 °C (10 mmHg)
:1.232
:135 °C (275 °F)
:246-644-8
:2810
:6.1,III
:GHS06
:GHS08
:GHS07
:Ambient
- Environmentally Preferable:
Frequently Bought Together