Order Entry
France
ContactUsLinkComponent
Tellurium, plasma standard solution, Te 10000 µg/ml, Specpure®
Tellurium, plasma standard solution, Te 10000 µg/ml, Specpure®
Catalog # 47187.AP
CAS Number:  
Tellurium, plasma standard solution, Te 10000 µg/ml, Specpure®
Catalog # 47187.AP
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    30% HCl
  • Pk:
    500 ml
  • MDL No.:
    MFCD00134062
  • Element:
    Tellurium
  • Symbol:
    Te

Specifications

About this item

Molecular Formula: Te in 30% HCl
Physical Form: Liquid
Odour: Acidic
UN No.: UN1789
MDL No.: MFCD00134062