Tellurium, plasma standard solution, Te 1000 µg/ml, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Te in 30% HCl
Physical Form: Liquid
Odour: Acidic
UN No.: UN1789
MDL No.: MFCD00134062
Specifications
- MDL No.:MFCD00134062
Specifications
Frequently Bought Together