Order Entry
France
ContactUsLinkComponent
Foot Sprays, Stokoderm® Foot Care
Foot Sprays, Stokoderm® Foot Care
Catalog # EVON32780_U
Supplier:  Deb-STOKO
undefined
Foot Sprays, Stokoderm® Foot Care
Catalog # EVON32780_U
Supplier:  Deb-STOKO
Specifications
  • Description:
    Foot spray, Stokoderm® Foot Care, 100 ml bottle
  • Pk:
    1
Specifications
About this item
This footspray helps to protect feet by preventing excessive perspiration, for instance when wearing closed in footwear, such as safety shoes, rubber boots etc. The alcoholic solution contains fragrances of essential oils and Hamamelis Extract, which toughens the horny layer of the feet.
  • Alcohol base provides Refresh™ing effect and helps provide effective protection against fungal skin infections
  • Essential oils provide protection against any unpleasant smells that may arise from the bacterial breakdown of sweat
  • Convenient spray form: Easy to use and apply to all areas
  • Also suitable for deodorising footwear
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904