Order Entry
France
ContactUsLinkComponent
Sodium nitrite ≥97.0%, granular, AR® ACS, Macron Fine Chemicals™
Oxidizing
Toxic
Environment
Sodium nitrite ≥97.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 7824-20
CAS Number:  
undefined
Oxidizing
Toxic
Environment
Sodium nitrite ≥97.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 7824-20
CAS Number:  

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay (NaNO₂):
    ≥97.0 %
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Chloride (Cl):
    ≤0.005 %
  • ACS - Sulfate (SO₄):
    ≤0.01 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • ACS - Calcium (Ca):
    ≤0.01 %
  • ACS - Potassium (K):
    ≤0.005 %
  • Appearance (white or yellowish white crystalline granules):

Specification Test Results

About this item

 :
 :69 g/mol
Merck Index:13,08721
Boiling Pt:320 °C (1013 hPa)
Melting Pt:284 °C
Density:2.168 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-555-9
UN:1500
ADR:5.1,III
 :GHS03
 :GHS06
 :GHS09
CAS Number:7632-00-0
Formula:NaNO₂
MDL Number:MFCD00011118