Order Entry
France
ContactUsLinkComponent
Multimodal chromatography columns, HiTrap™ Capto™ adhere ImpRes
Multimodal chromatography columns, HiTrap™ Capto™ adhere ImpRes
Supplier:  Cytiva
Certificates
undefined
Multimodal chromatography columns, HiTrap™ Capto™ adhere ImpRes
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 17-3715-10
  • 29-4004-64
  • Max. pressure:
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 12
  • 3 - 12