
3-Methyl-2-benzothiazolinonehydrazone hydrochloride, GR for analysis, Supelco®
Catalog # 1.04527.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Assay (argentometric):≥ 99%
- Identity (IR-spectrum):passes test
- pH-value (0.4 %, water):3 - 4
- Melting point:about 270°C
- Sulfated ash:≤ 0.1 %
- Loss on Drying (105°C):≤ 1.0 %
- Suitability for determination of aldehydes:passes test
- Bulk density:748 kg/m3
- Solubility:Slightly soluble at 20 °C
- Storage temperature:+15...+25 °C
- pH-value (0.4 %, water):2,5 - 4,4
- Loss on drying (105 °C):≤1,0 %
Specification Test Results
About this item
:
:215.70 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ100
UN:2811
:GHS06
CAS Number:4338-98-1
Formula:C₈H₁₀ClN₃S
MDL Number:MFCD00005816