Order Entry
France
ContactUsLinkComponent
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
Supplier:  Merck
Certificates
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    300 °C
  • Molar Mass:
    248.19 g/mol
  • HS Code:
    2917 20 00
  • Storage temperature:
    15...25 °C
  • Assay (acidimetric):
    ≥98%
  • Identity (IR):
    passes test

Specification Test Results

Related Information

Frequently Bought Together