Anti-RAG2 Rabbit Polyclonal Antibody
100 µg
Catalog # BSBTPB9793
Supplier: Boster Bio
Jump to:
Product Details & Documents
Rabbit IgG polyclonal antibody for V(D)J recombination-activating protein 2(RAG2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: RAG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
PB9793
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Isotype:IgG
- Host:Rabbit
- Cross adsorption:No
- Antigen name:Recombination activating gene 2
- Cat. No.:BSBTPB9793
- Reactivity:Human|Rat|Mouse
- Gene ID:5897
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:RAG2
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



