Order Entry
France
ContactUsLinkComponent
Crotonic anhydride cis- and trans-mixture for synthesis, Sigma-Aldrich®
Crotonic anhydride cis- and trans-mixture for synthesis, Sigma-Aldrich®
Catalog # 8.22002.0100
Supplier:  Merck
CAS Number:  
Crotonic anhydride cis- and trans-mixture for synthesis, Sigma-Aldrich®
Catalog # 8.22002.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    -20 °C
  • Molar Mass:
    154.17 g/mol
  • Boiling Point:
    250 °C
  • Vapor Pressure:
    1 hPa (25 °C)
  • Flash Point:
    113 °C
  • Refractive Index:
    1.4741 (20 °C, 589 nm)
  • Density:
    1.04 g/cm3 (20 °C)
  • Ignition Point:
    290 °C
  • Vapour pressure:
    1 hPa (25 °C)
  • Storage temperature:
    +15...+25 °C

Specification Test Results

About this item

 :
ADR:9,III
 :M-Clarity™ quality level = MQ200
 :154.17 g/mol
EINECS:210-807-1
UN:3082
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:623-68-7
Formula:C₈H₁₀O₃
MDL Number:MFCD00009286