Anti-RPUSD2 Rabbit Polyclonal Antibody
Product Details & Documents
RPUSD2, is a 545 amino acid member of the pseudouridine synthase rluA family and is encoded by a gene that is located on chromosome 15. Encoding more than 700 genes, chromosome 15 is made up of approximately 106 million base pairs and is about 3% of the human genome. Angelman and Prader-Willi syndromes are associated with loss of function or deletion of genes in the 15q11-q13 region. In the case of Angelman syndrome, this loss is due to inactivity of the maternal 15q11-q13 encoded UBE3A gene in the brain by either chromosomal deletion or mutation. In cases of Prader-Willi syndrome, there is a partial or complete deletion of this region from the paternal copy of chromosome 15. Tay-Sachs disease is a lethal disorder associated with mutations of the HEXA gene, which is encoded by chromosome 15. Marfan syndrome is associated with chromosome 15 through the FBN1 gene.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: RPUSD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-9626R
Datasheet
Specifications
- Size:100 μg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Purified by Protein A.
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Host:Rabbit
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Antigen name:RNA pseudouridylate synthase domain containing 2
- Cat. No.:BOSSBS-9626R
- Reactivity:Human|Rat|Mouse
- Gene ID:27079
- Antigen synonyms:C18B11|C15orf19
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:RPUSD2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.



