Anti-D.Aspartic acid Rabbit Polyclonal Antibody
Catalog # BOSSBS-8529R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:D.Aspartic acid
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Multiple
- Form:liquid
- Antigen synonyms:DL-Aminosuccinic acid|D Aspartic acid.|D-Aspartic acid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
D-Aspartic acid is the D-isomer of aspartic acid also known as aspartate which is one of the 20 natural proteinogenic amino acids, the building blocks of proteins. As its name indicates, aspartic acid is the carboxylic acid analog of asparagine. It is non-essential in mammals, and might serve as an excitatory neurotransmitter in the brain. It is also a metabolite in the urea cycle, and participates in gluconeogenesis.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: D.Aspartic acid
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others