Order Entry
France
ContactUsLinkComponent
Anti-FAS Rabbit Polyclonal Antibody
Anti-FAS Rabbit Polyclonal Antibody
Catalog # BOSSBS-5324R
Supplier:  Bioss
Anti-FAS Rabbit Polyclonal Antibody
Catalog # BOSSBS-5324R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fatty Acid Synthase
  • Antigen symbol:
    FAS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    355
  • Antigen synonyms:
    FASTM|APO-1|ALPS1A|TNFRSF6|CD95|APT1|FAS1
  • Modification:
    Tyr232
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Immunogen:
    210-240/335
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Receptor for TNFSF6/FASLG. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. FAS-mediated apoptosis may have a role in the induction of peripheral tolerance, in the antigen-stimulated suicide of mature T-cells, or both. The secreted isoforms 2 to 6 block apoptosis (in vitro).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CD95
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human