Anti-ATP2c1/2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-2434R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ATP dependent Ca(2+) pump PMR1
- Antigen symbol:ATP2c1/2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:27032, 9914
- Antigen synonyms:ATPase 2C1|ATPase 2C2|/Ca++ATPase|HUSSY 28|Secretory pathway Ca(2+)-ATPase 2.|ATP dependent Ca(2+) pump PMR1|Calcium-transporting ATPase type 2C member 2|KIAA1347|Calcium transporting ATPase type 2C member 1|PMR1L|ATP2C2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:315-352/919
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of calcium.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ATP2c1/2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat