Order Entry
France
Orders LinkContactUsLinkComponent
Anti-TFAP2B Rabbit Polyclonal Antibody

Anti-TFAP2B Rabbit Polyclonal Antibody

100 μg
Catalog # BOSSBS-12480R
Supplier:   Bioss
Anti-TFAP2B Rabbit Polyclonal Antibody
Catalog # BOSSBS-12480R
Supplier:  Bioss
Anti-TFAP2B Rabbit Polyclonal Antibody

100 μg

Price
649,00 €
Log in for availability
1 * 100 µl
Product Details & Documents

The AP2 proteins are normally expressed in ectodermally derived vertebrate tissues where they are necessary for normal growth and development. The factors have also been implicated in the control of cell proliferation, viral transformation, and oncogenesis. AP2 seems to play in important role in human breast cancer. AP2 alpha is the only AP2 protein required for early morphogenesis of the lens vesicle. AP2 beta appears to be required for normal face and limb development and for proper terminal differentiation and function of renal tubular epithelia.

  • WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: TFAP2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Documents

Specifications
  • Size:
    100 μg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Purified by Protein A.
  • Antibody type:
    Primary
  • Form:
    liquid
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Antigen name:
    Transcription Factor AP-2 beta
  • Cat. No.:
    BOSSBS-12480R
  • Reactivity:
    Human|Rat|Mouse
  • Gene ID:
    7021
  • Antigen synonyms:
    AP-2B|AP2-B
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    TFAP2B
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.