Order Entry
France
ContactUsLinkComponent
 
Anti-L-Citrulline Rabbit Polyclonal Antibody
  BOSSBS-10255R
 :  Bioss
Anti-L-Citrulline Rabbit Polyclonal Antibody
  BOSSBS-10255R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    L-Citrulline
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Multiple
  • Form:
    liquid
  • Antigen synonyms:
    S-2-Amino-5-ureidopentanoic Acid.|L-2-Amino-5-ureidovaleric Acid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

The amino acid Citrulline is required to detoxify the liver from ammonia, which is a waste product of the body from oxidation. Citrulline promotes energy and assists with the immune system. This unusual amino acid is formed in the urea cycle by the addition of carbon dioxide and ammonia to ornithine. It is then combined with aspartic acid to form arginosuccinic acid, which later is metabolized into the amino acid arginine.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: L-Citrulline
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others