Order Entry
France
Orders LinkContactUsLinkComponent
Phosphate buffer solution, ready to use, VWR®

Phosphate buffer solution, ready to use, VWR®

Phosphate buffer solution, 6 - 6,5, Plastic bottle, 2 L
Catalog # PRLSMC307202.2000
Supplier:   PROLABO CMP
Phosphate buffer solution, ready to use, VWR®
Catalog # PRLSMC307202.2000
Supplier:  PROLABO CMP
CAS Number:  
Phosphate buffer solution, ready to use, VWR®

Phosphate buffer solution, 6 - 6,5, Plastic bottle, 2 L

CAS Number:  
Price
74,70 €
Log in for availability
1 * 2 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2 L
  • Description:
    Phosphate buffer solution
  • pH value:
    6 - 6,5

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.