Order Entry
France
Orders LinkContactUsLinkComponent
Sample dividers
Sample dividers
Sample divider, stainless steel, 12 slots 6,35 mm (¼") width, volume 0,3 L
Catalog # ENDE1/4SSDSLOT
Supplier:   ENDECOTTS
Sample dividers
Catalog # ENDE1/4SSDSLOT
Supplier:  ENDECOTTS
Sample dividers
Sample dividers
Sample divider, stainless steel, 12 slots 6,35 mm (¼") width, volume 0,3 L
Price
2 430,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Endecotts hand held sample dividers will subdivide material samples into two smaller portions by a single pass. Further subdivisions can be attained by multiple passes. Each subdivision created retains the characteristics of the original sample.
  • Ideal for free flowing powders
  • Unit consists of divider plus 3 receiving boxes
  • Stainless steel with slot widths
  • Will subdivide material samples into two smaller portions by single pass
  • Each subdivision retains the characteristics of the original sample
  • Based on recommendations of BS 5309 and BS 3406/1
Specifications
  • Description:
    Sample divider, stainless steel, 12 slots 6,35 mm (¼") width, volume 0,3 L