Order Entry
France
ContactUsLinkComponent
Anti-ACC synthase 7 Rabbit Polyclonal Antibody
Anti-ACC synthase 7 Rabbit Polyclonal Antibody
Catalog # AGRIAS142774
Supplier:  Agrisera
undefined
Anti-ACC synthase 7 Rabbit Polyclonal Antibody
Catalog # AGRIAS142774
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ACC synthase 7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    A. thaliana
  • Western blot:
    Yes
  • UniProtKB:
    Q9STR4
  • Immunogen:
    KLH-conjugated synthetic peptide derived from Arabidopsis thaliana ACC synthase 7,
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

1-aminocyclopropane-1-carboxylate synthase (ACS) enzymes catalyze the conversion of S-adenosyl-L-methionine (SAM) into 1-aminocyclopropane-1-carboxylate (ACC), a direct precursor of ethylene. (EC:4.4.1.14). Synonymes: S-adenosyl-L-methionine methylthioadenosine-lyase 7.,1-aminocyclopropane-1-carboxylate synthase (ACS) enzymes catalyze the conversion of S-adenosyl-L-methionine (SAM) into 1-aminocyclopropane-1-carboxylate (ACC), a direct precursor of ethylene. (EC:4.4.1.14). Synonymes: S-adenosyl-L-methionine methylthioadenosine-lyase 7.

Type: Primary
Antigen: ACC synthase 7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana