Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
pH Calibration Buffers
VWR®, pH electrode maintenance kit
VWR®, pH electrode maintenance kit
VWR®, pH electrode maintenance kit
83609.600
:
VWR Chemicals
:
VWR®, pH electrode maintenance kit
83609.600
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
1 SET
Reference temperature:
20 °C
Colour:
Neutral
Description:
Buffers, electrolyte and cleaning solutions (100 ml)
Seq:
98
pH value:
4,00/7,00/10,00