Order Entry
France
ContactUsLinkComponent
 
Digital timer, one channel
  MOIN288CH
 :  MOINEAU
undefined
Digital timer, one channel
  MOIN288CH
 :  MOINEAU

 

  • Colour:
    White/red
  • Description:
    Digital timer, single channel
  • W×D×H:
    62×24×52 mm
  • Pk:
    1

 

 

Small laboratory timer.

  • Counts up and down
  • Timing capacity: 100 minutes
  • Alarm and memory function
  • Lightweight (only 40 g)
 : Supplied with magnet, stand and LR44 battery