Anti-Eph receptor A5 Antibody
BSBTPB9583
- Antibody type:Primary
- Antigen name:EPH receptor A5
- Antigen symbol:EPHA5
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2044
- Antigen synonyms:TYRO4|EHK-1|CEK7|EHK1|HEK7|EK7
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Ephrin type-A receptor 5(EPHA5) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: Eph receptor A5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human