Order Entry
France
ContactUsLinkComponent
Anti-Endothelin B receptor Rabbit Polyclonal Antibody
Anti-Endothelin B receptor Rabbit Polyclonal Antibody
Catalog # BSENR-1550-100
Supplier:  Biosensis
undefined
Anti-Endothelin B receptor Rabbit Polyclonal Antibody
Catalog # BSENR-1550-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Endothelin B Receptor
  • Antigen symbol:
    ETRB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Human Endothelin B receptor (299-313 aa).
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Antigen synonyms:
    ETRB,|ETrB
  • Immunogen:
    A peptide from human Endothelin B receptor (299-313 aa).
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

The Endothelin B receptor is a multi-pass membrane protein that belongs to the G-protein coupled receptor 1 family. It acts as a non-specific receptor for endothelin 1, 2, and 3.

Antibody detects ETRB at 51 kDa in WB in mouse fetus protein lysate. A weaker band at approx 75 kDa may also be observed (unknown).
Application Information:
WB. Suggested dilution of 1:1,000-1:5,000. Human Endothelin B receptor has a predicted length of 442 residues and MW of 50 kDa. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Endothelin B receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Endothelin B receptor (299-313 aa).
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat