Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microplates
HTS Microplates
Microplates, 96 well, flat bottom
Microplates, 96 well, flat bottom
Microplates, 96 well, flat bottom
ANICM09600P0
:
Anicrin
Microplates, 96 well, flat bottom
ANICM09600P0
:
Anicrin
No. of wells:
96
Colour:
Clear
Description:
96-well plate, flat-bottomed
Pk:
300