Order Entry
France
ContactUsLinkComponent
Human mononuclear cells from cord blood (hMNC-CB)
Human mononuclear cells from cord blood (hMNC-CB)
Catalog # C-14096
Supplier:  PROMOCELL
CAS Number:  
undefined
Human mononuclear cells from cord blood (hMNC-CB)
Catalog # C-14096
Supplier:  PROMOCELL
CAS Number:  

Specifications

  • Cell Format:
    Pelleted
  • Pk:
    2 mL
  • Description:
    Human mononuclear cells from cord blood (hMNC-CB), single donor, pellet, 1×106 cells in RNAlater®

Specifications

About this item

Human mononuclear cells from cord blood (hMNC-CB) are isolated from umbilical cord blood, which has a much higher content of primitive progenitor cells compared to adult peripheral blood.

Applications include: Stem cell research, transplantation basic research, hematologic and immunologic studies, oncology research, virology and infection biology.