Human bronchial epithelial cells (HBEpC)
Catalog # C-14063
Supplier: PROMOCELL
CAS Number:
Specifications
- Cell Format:Pelleted
- Pk:2 mL
- Description:Human bronchial epithelial cells (HBEpC) pellet, 1×10⁶ cells in RNAlater®
Specifications
About this item
Human bronchial epithelial cells (HBEpC) are isolated from the surface epithelium of human bronchi and stain positive for cytokeratin. The respiratory epithelia are responsible for the lubrication of the lungs, the maintenance of humidity, and the cleaning of the respiratory tract. They are an important target for drugs, toxins, and carcinogens. In addition, they are involved in many diseases such as cystic fibrosis. Consequently, HBEpC are useful for investigating the function and pathology of the respiratory system.
- Cytokeratin positive
- Routinely morphology checked
- Cryopreserved at second passage
This product can be used in the following applications. Drug uptake studies, Toxicological research, Wound healing research, Oncology, Cell differentiation studies, Respiratory disease research.
Ordering information: Human Bronchial Epithelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.