Anti-CHRNA9 Mouse Monoclonal Antibody [clone: 8E4]
Catalog # ABNOH00055584-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cholinergic receptor, nicotinic, alpha 9 (neuronal)
- Antigen symbol:CHRNA9
- Clonality:Monoclonal
- Clone:8E4
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:55584
- Antigen synonyms:HSA243342|NACHRA9
- Amino acid number:139 to 221
- Storage buffer:1x PBS, pH 7,4
- Sequence:YDGLITWDAPAITKSSCVVDVTYFPFDNQQCNLTFGSWTYNGNQVDIFNALDSGDLSDFIEDVEWEVHGMPAVKNVISYGCCS
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CHRNA9 (NP_060051.2, 139 a.a. ~ 221 a.a) partial recombinant protein with GST tag.
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant CHRNA9.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CHRNA9
Clonality: Monoclonal
Clone: 8E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: