Order Entry
France
ContactUsLinkComponent
 
Anti-ZBTB20 Mouse Monoclonal Antibody [clone: 1F3]
  ABNOH00026137-M01
 :  Abnova
Anti-ZBTB20 Mouse Monoclonal Antibody [clone: 1F3]
  ABNOH00026137-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger and BTB Domain Containing 20
  • Antigen symbol:
    ZBTB20
  • Clonality:
    Monoclonal
  • Clone:
    1F3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    26137
  • Antigen synonyms:
    ODA-8S|ZNF288|DPZF|PRIMS|HOF
  • Amino acid number:
    451 to 542
  • Storage buffer:
    1x PBS, pH 7,4
  • Sequence:
    FLFSLPQPLAGQQTQFVTVSQPGLSTFTAQLPAPQPLASSAGHSTASGQGEKKPYECTLCNKTFTAKQNYVKHMFVHTGEKPHQCSICWRSF
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    ZBTB20 (NP_056457, 451 a.a. ~ 542 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against a partial recombinant ZBTB20.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: ZBTB20
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human