Anti-ADK Mouse Monoclonal Antibody [clone: AT4F8]
Catalog # ABNOMAB10016
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Adenosine Kinase
- Antigen symbol:ADK
- Clonality:Monoclonal
- Clone:AT4F8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:132
- Antigen synonyms:Adenosine 5'-phosphotransferase|AK
- Amino acid number:22 to 362
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 22-362 of human ADK.
- Purification:Protein G purification
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant ADK.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: ADK
Clonality: Monoclonal
Clone: AT4F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human