Anti-MST1R Mouse Monoclonal Antibody [clone: 1B5A9]
Catalog # ABNOMAB10354
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Macrophage Stimulating 1 Receptor
- Antigen symbol:MST1R
- Clonality:Monoclonal
- Clone:1B5A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:4486
- Antigen synonyms:soluble R|RON variant E2E3|MST1R variant RON30|PTK8|macrophage-stimulating protein receptor|MST1R variant RON62|soluble RON variant 1|c-met-related tyrosine kinase|CDw136|PTK8 protein tyrosine kinase 8|MSP receptor|p185-Ron|soluble RON variant 3|RON|soluble RON variant 2|CD136
- Amino acid number:210 to 320
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 210-320 of human MST1R.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant MST1R.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: MST1R
Clonality: Monoclonal
Clone: 1B5A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human