Order Entry
France
ContactUsLinkComponent
Anti-S100 A1 Mouse Monoclonal Antibody [clone: 2C8B8,2C8E8,3C8D8]
Anti-S100 A1 Mouse Monoclonal Antibody [clone: 2C8B8,2C8E8,3C8D8]
Catalog # ABNOMAB10365
Supplier:  Abnova
Anti-S100 A1 Mouse Monoclonal Antibody [clone: 2C8B8,2C8E8,3C8D8]
Catalog # ABNOMAB10365
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    S100 Calcium Binding Protein A1
  • Antigen symbol:
    S100A1
  • Clonality:
    Monoclonal
  • Clone:
    2C8B8,2C8E8,3C8D8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites
  • Gene ID:
    6271
  • Antigen synonyms:
    S100|S100-alpha|S100A
  • Storage buffer:
    0,03% sodium azide
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human S100A1.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against recombinant S100A1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: S100 A1
Clonality: Monoclonal
Clone: 2C8B8,2C8E8,3C8D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human