Anti-TRDMT1 Rabbit Polyclonal Antibody
ABNOPAB0161
: Abnova
- Antibody type:Primary
- Antigen name:tRNA Aspartic Acid Methyltransferase 1
- Antigen symbol:TRDMT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:13434
- Antigen synonyms:DMNT2|DNMT2|MHSAIIP|PUMET|RNMT1
- Amino acid number:39 to 53 and 361 to 376
- Storage buffer:PBS (0,05% BSA, 0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A mixture of synthetic peptides corresponding to amino acids 39-53 and 361-376 of mouse Dnmt2.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of Trdmt1.
Type: Primary
Antigen: TRDMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse