Order Entry
France
ContactUsLinkComponent
Anti-PRKAB1 Rabbit Polyclonal Antibody
Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB16984
Supplier:  Abnova
Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB16984
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase, AMP-activated Non-catalytic Subunit Beta 1
  • Antigen symbol:
    PRKAB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5564
  • Antigen synonyms:
    kinase AMPK-beta1|AMPK-beta1|AMPK beta-1 chain|5'-AMP-activated protein kinase beta-1 subunit|PRKAB1|AAKB1
  • Storage buffer:
    20 mM PBS, 0,15 M NaCl, pH 7,2 (0,01% sodium azide)
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to residues surrounding S182 of human PRKAB1.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of PRKAB1.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: PRKAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat